Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium 2,4-pentanedionate hydrate, typically 98+% (metals basis)

$310.93 - $560.30

Chemical Identifiers

Chemical Name or Material Magnesium 2,4-pentanedionate hydrate
CAS 14024-56-7
Formula Weight 222.52 (Anhydrous)
MDL Number MFCD00150168
Synonym Bis(acetylacetonato)magnesium; Bis(2,4-pentanedionato)magnesium
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1253209

 
alfa aesar™
1253209
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1253218

 
alfa aesar™
1253218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1253230

 
alfa aesar™
1253230
250g Each for $310.93

AA1253236

 
alfa aesar™
1253236
500g Each for $560.30
Description & Specifications

Specifications

Assay ≥98% (typically)
Formula Weight 222.52 (Anhydrous)
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Magnesium 2,4-pentanedionate hydrate
Assay Percent Notes (metals basis)
MDL Number MFCD00150168
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Color White
Melting Point 260°C (dec.)
Quantity 10g
Physical Form Powder
CAS 14024-56-7
Synonym Bis(acetylacetonato)magnesium; Bis(2,4-pentanedionato)magnesium
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.