missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium 2,4-pentanedionate hydrate, typically 98+% (metals basis)

Manufacturer:  Alfa Aesar™ 1253236

 View more versions of this product

Catalog No. AA1253236



Specifications

Specifications

Magnesium 2,4-pentanedionate hydrate
260°C (decomposition)
14024-56-7
C10H14MgO4
4-4-oxopent-2-en-2-yl oxy magnesio oxy pent-3-en-2-one
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].[Mg+2]
222.523
222.52 (Anhydrous)
≥98% (typically)
White
500g
(metals basis)
MFCD00150168
AKTIAGQCYPCKFX-SYWGCQIGSA-L
magnesium;(E)-4-oxopent-2-en-2-olate
25113476
Powder

Chemical Identifiers

14024-56-7
222.523
AKTIAGQCYPCKFX-SYWGCQIGSA-L
25113476
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].[Mg+2]
C10H14MgO4
MFCD00150168
4-4-oxopent-2-en-2-yl oxy magnesio oxy pent-3-en-2-one
magnesium;(E)-4-oxopent-2-en-2-olate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents