Please login to your online account to display your discounted pricing

Magnesium, 1000μg/mL (0.10% w/v), BAKER INSTRA-ANALYZED™ Reagent, J.T.Baker™

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

$118.18 - $410.89

Chemical Identifiers

Chemical Name or Material Magnesium, 1000μg/mL (0.10% w/v)
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Molecular Formula Mg
Formula Weight 24.31
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003253

 
avantor performance
576704
Poly Bottle 100mL Each for $410.89

02003254

 
avantor performance
645704
Narrow Mouth Poly Bottle 150mL Each for $118.18
Description & Specifications

Specifications

Formula Weight 24.31
Molecular Formula Mg
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Quantity 100mL
Specific Gravity 1L = 1.01kg
Packaging Poly Bottle
Chemical Name or Material Magnesium, 1000μg/mL (0.10% w/v)