Magleads
Make working electrical circuits instantly.
Manufacturer:  Fisher Science Education 61552
 View more versions of this product
Catalog No. S94395
Alert:
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Description
Description
  • Contain tiny, yet powerful magnets at each end
  • Self-connect to battery case and bulb holder connector to create a circuit
  • 4mm magnets work anywhere standard clips are used
  • Can be daisy chained together to extend length
  • Pack of 10
Specifications
Specifications
Black
0.4 cm
11.8 in.
Electrical Circuit Lead
0.15 in.
Circuit creation, use in place of standard clips, can be daisy chained to extend length
30 cm