Please login to your online account to display your discounted pricing
Magleads
Make working electrical circuits instantly.
$31.05 - $36.12
Specifications
Diameter (English) Inside 0.15 in.
Diameter (Metric) Inside 0.4cm
Product Type Electrical Circuit Lead
For Use With (Application) Circuit creation, use in place of standard clips, can be daisy chained to extend length
View More Specs
 Alert
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Products
Catalog Number Mfr. No. Color Length (English) Length (Metric) Price Quantity    
S94396
fisher science education
61553
Red 11.8 in. 30cm Each for $36.12
S94395
fisher science education
61552
Black 11.8 in. 30cm Each for $36.12
S94394
fisher science education
61551
Red 5.9 in. 15cm Each for $31.05
Description & Specifications
Specifications
Diameter (English) Inside 0.15 in.
Diameter (Metric) Inside 0.4cm
Product Type Electrical Circuit Lead
For Use With (Application) Circuit creation, use in place of standard clips, can be daisy chained to extend length
  • Contain tiny, yet powerful magnets at each end
  • Self-connect to battery case and bulb holder connector to create a circuit
  • 4mm magnets work anywhere standard clips are used
  • Can be daisy chained together to extend length
  • Pack of 10