Thermo Scientific™ MagJET Genomic DNA Kit
Purify genomic DNA from cell culture, tissue samples, bacteria, and yeast with a magnetic bead separation method amenable to high-throughput workflows.
$289.00 - $950.00
Specifications
Storage Requirements Ambient
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
FERK2721 Thermo Scientific™
K2721
96 Preps Each for $289.00
FERK2722 Thermo Scientific™
K2722
4 x 96 Preps Each for $950.00
Specifications
Description
MagJET Genomic DNA Kit is designed for fast and efficient purification of genomic DNA from tissue and cell cultures, bacteria and yeast, as well as from human body samples, such as buccal and urogenital swabs, urine, saliva and hair follicles.
The kit utilizes paramagnetic bead technology enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users.
The resulting high quality DNA is free of proteins, nucleases and other contaminants or inhibitors, can be used in a wide range of downstream applications such as PCR, qPCR and other enzymatic reactions.
Highlights
  • Versatile -suitable for gDNA purification from variety of biological samples, including gDNA from eukaryotic cells, animal tissues, yeast and bacteria, as well as from human body samples, such as buccal and urogenital swabs, urine, saliva and hair follicles
  • Efficient -high yields of high molecular weight genomic DNA
  • High quality -isolates high integrity gDNA, free of inhibitors and RNA contamination
  • Flexible -easily adjustable for high throughput automation platforms and manual processing
Description & Specifications
Specifications
Ambient