Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MAGE-1 Ab-6, Mouse Monoclonal Antibody

Ideal for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ MAGE-1 Ab-6, Mouse Monoclonal Antibody provides accurate, reproducible results.

$262.04 - $1,227.55

Specifications

Antigen MAGE-1 Ab-6
Clone 6C1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS1836R7

 
thermo scientific™ lab vision™
MS1836R7
7mL Each for $634.82

MS1836S

 
thermo scientific™ lab vision™
MS1836S
1mL Each for $1,227.55

MS1836S0

 
thermo scientific™ lab vision™
MS1836S0
100µL Each for $262.04

MS1836S1

 
thermo scientific™ lab vision™
MS1836S1
500µL Each for $845.33
Description & Specifications

Specifications

Antigen MAGE-1 Ab-6
Clone 6C1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Type Primary
Immunogen Full length recombinant human MAGE-1 protein
Isotype IgG2a

Ideal for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ MAGE-1 Ab-6, Mouse Monoclonal Antibody provides accurate, reproducible results.

Human malignant neoplasms carry rejection antigens that are recognized by the patients' autologous, tumor-directed and specific, cytolytic, CD8+ T lymphocyte clones (CTL). An important group of antigens is coded by the MAGE family of genes. It was identified that melanomas and primary glial brain tumors express common melanoma associated antigens (MAAs). Because MAGE-1 is expressed on a significant proportion of human neoplasms of various histological types (melanoma, brain tumors of glial origin, neuroblastoma, non-small cell lung cancer, breast, gastric, colorectal, ovarian, renal cell carcinomas) and not on normal tissues, the encoded antigen may serve as a marker of early detection and target for cancer immunotherapy

Host Species: Mouse

Clone: 6C1

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Full-length recombinant human MAGE-1 protein

Molecular Weight: 42-46kDa

Positive Control: Hepatocellular CA, normal testis

Cellular Localization: Cytoplasmic, occasionally nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO