Please login to your online account to display your discounted pricing

Thermo Scientific MAGE-1 Ab-4, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in experiments with the Thermo Scientific™ MAGE-1 Ab-4, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific  MS1067PABX

 View more versions of this product

Catalog No. MS1067PABX

  • / Each
 

Description & Specifications

Specifications

Quantity 200µL
Isotype IgG2a kappa
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Rat
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Partially purified, full length recombinant human MAGE-1 protein
Host Species Mouse
Clone MA454
Regulatory Status RUO
Format Concentrated
Antigen MAGE-1 Ab-4
Type Primary
Concentration 1mg/mL
Formulation Ab Without BSA/Azide (Frozen)
Classification Monoclonal
Conjugate Unlabeled
Research Discipline Cancer and Tumor Biology

Ensure accurate, reproducible results in experiments with the Thermo Scientific™ MAGE-1 Ab-4, Mouse Monoclonal Antibody.

Human malignant neoplasms carry rejection antigens that are recognized by the patients' autologous, tumor directed and specific, cytolytic, CD8+ T lymphocyte clones (CTL). An important group of antigens is coded by the MAGE family of genes. It was identified that melanomas and primary glial brain tumors express common melanoma associated antigens (MAAs). Because MAGE-1 is expressed on a significant proportion of human neoplasms of various histological types (melanoma, brain tumors of glial origin, neuroblastoma, non-small cell lung cancer, breast, gastric, colorectal, ovarian, renal cell carcinomas) and not on normal tissues, the encoded antigen may serve as a marker of early detection and target for cancer immunotherapy

Host Species: Mouse

Clone: MA454

Isotype: IgG1/κ

Species Reactivity: Human, Dog and Rat. Others not known.

Immunogen: Partially purified, full-length recombinant human MAGE-1 protein

Molecular Weight: 42-46kDa

Positive Control: Colo32 cells germ cells in testis

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO