Please login to your online account to display your discounted pricing

Alfa Aesar™ Magainin II

For cellular and molecular biology applications

Specifications

Product Type Magainin II
Molecular Weight 2466.93
Storage Requirements -30° to -10°C
Sequence Gly-Ile-Gly-Lys-Phe-Leu-His-Ser-Ala-Lys-Lys-Phe-Gly-Lys-Ala-Phe-Val-Gly-Glu-Ile-Met-Asn-Ser
For Use With (Application) Peptide with potent antibiotic properties
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ61172EXC

 
alfa aesar™
J61172EXC
2.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ61172EXD

 
alfa aesar™
J61172EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ61172MCR

 
alfa aesar™
J61172MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Magainin II
Molecular Weight 2466.93
Storage Requirements -30° to -10°C
Sequence Gly-Ile-Gly-Lys-Phe-Leu-His-Ser-Ala-Lys-Lys-Phe-Gly-Lys-Ala-Phe-Val-Gly-Glu-Ile-Met-Asn-Ser
For Use With (Application) Peptide with potent antibiotic properties
Format Powder
Regulatory Status RUO
Purification Method HPLC