missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
LaMotte™ Macro Mania Expansion Kit
Expansion Kit for use with Macro Mania game (5942). Classroom adventure introduces the use of stream macroinvertebrates to determine water quality.
Supplier: LaMotte™ 5943
Includes: Expansion kit includes: cards (3 decks) and sorting sheets (3).
Description
- Students learn about the influence of land use on water quality and the practice of macroinvertebrate sampling to measure water quality
- Contains background information and step-by-step instructions for activities and extensions
- Integrates math, public speaking, writing and art
Order Info
Class size: six teams. Expansion kit adds three more teams.
Specifications
Game | |
Macro Mania Expansion Kit | |
Introduce the use of stream macroinvertebrates to determine Water quality. Lecture material sets the scene for the activity |
For use with LaMotte Macro Mania game (5942). | |
Expansion kit includes: cards (3 decks) and sorting sheets (3) | |
Science |