Please login to your online account to display your discounted pricing

Macro Mania

Classroom adventure introduces the use of stream macroinvertebrates to determine water quality.

$30.75 - $80.00

Specifications

Category Science
Format Game
For Use With (Application) Introduce the use of stream macroinvertebrates to determine Water quality. Lecture material sets the scene for the activity
Includes Cards (6 decks), sorting sheets (6), instruction manual and colorful Macro Mania classroom poster. Expansion kit includes: cards (3 decks) and sorting sheets (3)
View More Specs
Includes:  Cards (six decks), sorting sheets (6), instruction manual, and colorful Macro Mania classroom poster. Expansion kit includes: cards (three decks) and sorting sheets (3).
Products
Catalog Number Mfr. No. Product Type Price Quantity    

S85018

 
fisher science education
5942
Macro Mania Adventure Each for $80.00

S85018EK

 
fisher science education
5943
Macro Mania Kit Each for $30.75
Description & Specifications

Specifications

Category Science
Format Game
For Use With (Application) Introduce the use of stream macroinvertebrates to determine Water quality. Lecture material sets the scene for the activity
Includes Cards (6 decks), sorting sheets (6), instruction manual and colorful Macro Mania classroom poster. Expansion kit includes: cards (3 decks) and sorting sheets (3)

  • Students learn about the influence of land use on water quality and the practice of macroinvertebrate sampling to measure water quality
  • Contains background information and step-by-step instructions for activities and extensions
  • Integrates math, public speaking, writing and art

Class size: six teams. Expansion kit adds three more teams.