Please login to your online account to display your discounted pricing

GE Healthcare MabSelect Sure™ Chromatography Media

MabSelect™ SuRe™ is a protein A affinity medium for capturing MAbs at process scale with an alkali-tolerant rProtein A ligand that allows the for cost-effective sanitization protocols

$6,601.76 - $6,601.76

Specifications

Binding Capacity 30mg Human lgG/mL at 2.4 min. residence time
Particle Size 85μm
Product Type MabSelect SuRe media
Flow Rate <500cm/Hour
For Use With (Application) For use of rigorous and cost-effective CIP and sanitization protocols, manufacture of monoclonal antibodies
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

45-002-989

 
ge healthcare
17543802
200mL Each for $6,601.76
Description & Specifications

Specifications

Binding Capacity 30mg Human lgG/mL at 2.4 min. residence time
Particle Size 85μm
Product Type MabSelect SuRe media
Flow Rate <500cm/Hour
For Use With (Application) For use of rigorous and cost-effective CIP and sanitization protocols, manufacture of monoclonal antibodies
Height (English) Bed 7.87 in.
Height (Metric) Bed 20cm
Includes Certificate of Analysis
Ligand Type Alkali-stabilized Protein A-derived (E. coli)
pH Range 3 to 12
Stability Stable in aqueous Buffers commonly used in Protein A Chromatography
Storage Requirements 2 to 8°C

Has an alkali-tolerant rProteain A ligand designed to enhance protease resistance to produce monoclonal antibodies

  • Withstands rigorous CIP and sanitization procedures with 0.1 to 0.5 M NaOH
  • Design gives enhanced protease resistance resulting in lower ligand leakage
  • Generic elution profile for different monoclonal antibodies enables platform approach to purification
  • High-flow agarose matrix allows the processing of large feed volumes
  • Used in the commercial manufacture of monoclonal antibodies