Cytiva MabSelect PrismA Chromatography Resin PROMO
MabSelect PrismA Chromatography Resin is a Protein A Chromatography Resin. MabSelect PrismA has been improved with an optimized high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing future demands in mAb processing to be met.
Manufacturer:  Cytiva 29822171
Catalog No. 09928073LR2
Description
Description
  • Enhanced dynamic binding capacity allows high mass throughput of processed mAb per resin volume unit.
  • Excellent alkaline stability enables efficient cleaning and sanitization using 0.5–1.0 M NaOH for improved process economy, bioburden control, and robustness.
  • Covered by a comprehensive security of supply program, including dual sources of the agarose base matrix and protein A ligand.
  • The PrismA protein A ligand is also available in HiTrap Fibro™ and HiScreen Fibro™ PrismA chromatography units for ultrafast purification of mAbs.
Specifications
Specifications
13 mL
0.59 in.
Highly cross-linked agarose, spherical.
300 cm/hr.
25 mL
Chromatography Resin
80 mg human IgG/mL resin at 6 min. residence time. 65 mg human IgG/mL resin at 4 min. residence time.
1.5 cm
2 to 8°C, 20% ethanol
60 μm
Antibody affinity chromatography
Alkaline-stabilized protein A-derived (E.coli).