Please login to your online account to display your discounted pricing

Thermo Scientific™ MAbPac™ SEC-1 Size Exclusion LC Columns


Use these SEC columns for monoclonal antibody (mAb) analysis, including the high-resolution separation of monomers, aggregates and fragments.

Manufacturer: thermo scientific™  074696

 View more versions of this product

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Catalog No. DX074696

  • / Each
 

Description & Specifications

Specifications

Column Type Size Exclusion
For Use With ----
Column Chemistry Proprietary Diol
USP Type L59
Max. Pressure 1800psi
Packing Material Spherical, fully porous ultrapure silica
Particle Size 5μm
Temperature 30°C
Pore Size 300Å
pH 2.5 to 7.5
Stationary Phase Diol
Solvents 100% Organic Solvents
Length (Metric) 300mm
Material PEEK
Column Format Analytical Column
Diameter (Metric) 4mm

Use Thermo Scientific™ MAbPac™ SEC-1 Size Exclusion Columns for high-resolution separation of monoclonal antibody (mAb) analysis, including the monomers, aggregates and Fab and Fc fragments resulting from proteolysis. These size-exclusion chromatography (SEC) columns provide superior performance, even under nondenaturing conditions, and in high- and low-salt mobile phases and volatile eluents.

High-Performance SEC Columns for MAb Analysis
  • Hydrophilic bonded layer minimizes undesired interactions between the biomolecules and the stationary phase
  • Nonmetallic and biocompatible PEEK housing eliminates metal contamination from the column hardware
  • Stable surface bonding leads to low column bleed and compatibility with MS, ELSD and Thermo Scientific™ Dionex™ Corona™ CAD detection
  • Rugged column packing provides reliability, reproducibility and long column life
  • Columns are available in 5μm particle size and 150 and 300mm lengths, with an average pore diameter of 300Å