Thermo Scientific™ MAbPac™ RP LC Columns
Get higher resolution using reversed-phase conditions when separating and characterizing monoclonal antibodies by HPLC and LC/MS.
Manufacturer:  Thermo Scientific™ 88649
 View more versions of this product
Catalog No. 08-864-9
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Use the new Thermo Scientific™ MAbPac™ RP columns for high-performance reversed-phase (RP) HPLC and LC/MS characterization of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins. These columns have excellent resolution, long column lifetime, and low carry-over. The surface chemistry, particle size, and pore size, have been optimized for high resolution separations.
Description & Specifications
Specifications
Guard
10mm
DVB
0 - 14
<110°C
2.1mm
4,000psi
4μm
1500Å