Learn More
m-Xylylene dibromide, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2495918
Description
It is used in preparation of four novel dinuclear half-titanocenes with meta- and orthoxylene bridges as new metallocenes. It was used in the synthesis of dissymmetrical bis-macro cyclic salt.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in preparation of four novel dinuclear half-titanocenes with meta- and orthoxylene bridges as new metallocenes. It was used in the synthesis of dissymmetrical bis-macro cyclic salt.
Solubility
Insoluble in water.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents, active metals, bases.
Specifications
m-Xylylene dibromide | |
74°C to 76°C | |
MFCD00000178 | |
UN3261 | |
1,3-bis bromomethyl benzene, m-xylylene dibromide, alpha,alpha'-dibromo-m-xylene, benzene, 1,3-bis bromomethyl, m-bis bromomethyl benzene, m-xylylene bromide, ccris 1777, m-xylyene dibromide, m-xylene, .alpha.,.alpha.'-dibromo, m-.alpha.,.alpha.'-dibromoxylene | |
OXHOPZLBSSTTBU-UHFFFAOYSA-N | |
1,3-bis(bromomethyl)benzene | |
69373 | |
97% |
626-15-3 | |
C8H8Br2 | |
50 g | |
971085 | |
Insoluble in water. | |
C1=CC(=CC(=C1)CBr)CBr | |
263.96 | |
263.96 |
Chemical Identifiers
626-15-3 | |
263.96 | |
OXHOPZLBSSTTBU-UHFFFAOYSA-N | |
69373 | |
C1=CC(=CC(=C1)CBr)CBr |
C8H8Br2 | |
MFCD00000178 | |
1,3-bis bromomethyl benzene, m-xylylene dibromide, alpha,alpha'-dibromo-m-xylene, benzene, 1,3-bis bromomethyl, m-bis bromomethyl benzene, m-xylylene bromide, ccris 1777, m-xylyene dibromide, m-xylene, .alpha.,.alpha.'-dibromo, m-.alpha.,.alpha.'-dibromoxylene | |
1,3-bis(bromomethyl)benzene |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 210-931-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only