Learn More
m-Xylene, 99%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 158930010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
m-Xylene | |
108-38-3 | |
0.8640g/mL | |
1 L | |
1.4960 to 1.4980 | |
0.864 | |
C6H4(CH3)2 | |
0.62 mPa.s (20°C) | |
Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, ether and most organic, solvents | |
CC1=CC(C)=CC=C1 | |
106.17 | |
CHEBI:28488 | |
99% |
-48.0°C | |
clear colorless liquid | |
139.0°C | |
25°C | |
99% | |
C8H10 | |
MFCD00008536 | |
m-xylene, 1,3-dimethylbenzene, m-xylol, m-dimethylbenzene, meta-xylene, m-methyltoluene, 3-xylene, benzene, 1,3-dimethyl, 1,3-dimethylbenzol, santosol 150 | |
IVSZLXZYQVIEFR-UHFFFAOYSA-N | |
1,3-xylene | |
7929 | |
106.17 | |
Extra Dry |
Chemical Identifiers
108-38-3 | |
106.17 | |
IVSZLXZYQVIEFR-UHFFFAOYSA-N | |
7929 | |
1,3-xylene |
C8H10 | |
MFCD00008536 | |
m-xylene, 1,3-dimethylbenzene, m-xylol, m-dimethylbenzene, meta-xylene, m-methyltoluene, 3-xylene, benzene, 1,3-dimethyl, 1,3-dimethylbenzol, santosol 150 | |
CHEBI:28488 | |
CC1=CC(C)=CC=C1 |
Safety and Handling
GHS H Statement:
Causes skin irritation
Harmful in contact with skin
Harmful if inhaled
Flammable liquid and vapor
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
EINECSNumber : 203-576-3
RUO – Research Use Only