missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

m-Xylene, 99%, Extra Dry, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC158931000

 View more versions of this product

Catalog No. AC158931000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

m-Xylene
clear colorless liquid
C8H10
MFCD00008536
Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, ether and most organic, solvents
CC1=CC(=CC=C1)C
106.168
CHEBI:28488
99%
0.8640g/mL
Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, ether and most organic, solvents
25°C
139.0°C
100mL
108-38-3
99%
C6H4(CH3)2
1,3-dimethylbenzene
IVSZLXZYQVIEFR-UHFFFAOYSA-N
1,3-xylene
7929
106.17
Extra Dry
1.4960 to 1.4980
0.864
0.62 mPa.s (20°C)
-48.0°C

Chemical Identifiers

108-38-3
106.168
IVSZLXZYQVIEFR-UHFFFAOYSA-N
7929
1,3-xylene
C8H10
MFCD00008536
1,3-dimethylbenzene
CHEBI:28488
CC1=CC(=CC=C1)C
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation
Harmful in contact with skin
Harmful if inhaled
Flammable liquid and vapor

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 203-576-3

SDS Documents