Argos Technologies M-Vac Aspirating System
Argos M-Vac and M-Vac Jr. systems are used to collect a variety of liquids and supernatants including tissue and cell culture media, biological immunology reagents, or any liquid chemical waste that uses a standard in-house vacuum. 
Manufacturer:  Argos Technologies EV446
 View more versions of this product
Catalog No. 03-391-211
Description
Description
Ideal for use with critical or dangerous liquid Autoclavable at 121°C for 20 minutes Withstands 700mm Hg
Specifications
Specifications
White
2L bottle with tubing, connected to in-house vacuum
Stainless Steel
EV446
M-Vac Jr. Stainless Steel Stand (2-L bottle sold separately) 1/EA
One 2L bottle
Bottle Holder
Documents