Please login to your online account to display your discounted pricing

Alfa Aesar™ m-Tolylurea, 98%

$28.64 - $198.89

Chemical Identifiers

Chemical Name or Material m-Tolylurea
CAS 63-99-0
Molecular Formula C8H10N2O
Formula Weight 150.18
MDL Number MFCD00025425
Beilstein 2639488
Synonym 3-Methylphenylurea
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2074906

  SDS
alfa aesar™
B2074906
5g Each for $28.64

AAB2074914

  SDS
alfa aesar™
B2074914
25g Each for $59.38

AAB2074922

  SDS
alfa aesar™
B2074922
100g Each for $198.89
Description & Specifications

Specifications

Formula Weight 150.18
Chemical Name or Material m-Tolylurea
Assay 98%
Molecular Formula C8H10N2O
MDL Number MFCD00025425
CAS 63-99-0
Beilstein 2639488
Synonym 3-Methylphenylurea
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 139° to 142°C
Quantity 5g
RTECS Number YU4025000
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.