Please login to your online account to display your discounted pricing

Alfa Aesar™ m-Tolylhydrazine, 97%

Chemical Identifiers

Chemical Name or Material m-Tolylhydrazine
CAS 536-89-0
Molecular Formula C7H10N2
Formula Weight 122.17
MDL Number MFCD00047815
UN Number UN2810
Beilstein 742264
Merck Index 14,9542
Synonym 3-Methylphenylhydrazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1195804

  SDS
alfa aesar™
L1195804
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1195809

  SDS
alfa aesar™
L1195809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 122.17
Assay 97%
Molecular Formula C7H10N2
MDL Number MFCD00047815
CAS 536-89-0
Beilstein 742264
Synonym 3-Methylphenylhydrazine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material m-Tolylhydrazine
UN Number UN2810
Merck Index 14,9542
Boiling Point 132° to 134°C (16mmHg)
Quantity 2g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

Applications
m-Tolylhydrazine is used as pharmaceutical intermediate. It reacts with cyclohexanone and obtain the 7-methyl-1,2,3,4-tetrahydro-carbazole.

Solubility
Slightly soluble in water.

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.