missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ m-Tolylhydrazine, 97%

Manufacturer:  Alfa Aesar™ L1195809

 View more versions of this product

Catalog No. AAL1195809



Description

Description

Applications
m-Tolylhydrazine is used as pharmaceutical intermediate. It reacts with cyclohexanone and obtain the 7-methyl-1,2,3,4-tetrahydro-carbazole.

Solubility
Slightly soluble in water.

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

m-Tolylhydrazine
10g
C7H10N2
UN2810
14,9542
GPTOGZLZMLJZCV-UHFFFAOYSA-N
(3-methylphenyl)hydrazine
68308
97%
132°C to 134°C (16mmHg)
536-89-0
MFCD00047815
742264
m-tolylhydrazine, 3-methylphenyl hydrazine, hydrazine, 3-methylphenyl, m-tolylhydrazin, m-toluidine amine, m-tolyl hydrazine, m-tolyl-hydrazine, 1-m-tolylhydrazine, 3-methylphenylhydrazinehydrochloride, acmc-20akdk
CC1=CC(=CC=C1)NN
122.171
122.17

Chemical Identifiers

536-89-0
122.171
GPTOGZLZMLJZCV-UHFFFAOYSA-N
68308
CC1=CC(=CC=C1)NN
C7H10N2
MFCD00047815
m-tolylhydrazine, 3-methylphenyl hydrazine, hydrazine, 3-methylphenyl, m-tolylhydrazin, m-toluidine amine, m-tolyl hydrazine, m-tolyl-hydrazine, 1-m-tolylhydrazine, 3-methylphenylhydrazinehydrochloride, acmc-20akdk
(3-methylphenyl)hydrazine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

Documents