missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ m-Tolylacetonitrile, 98%

$271.42 - $271.42

Chemical Identifiers

CAS 2947-60-6
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00001914
InChI Key WOJADIOTNFDWNQ-UHFFFAOYSA-N
Synonym 2-3-methylphenyl acetonitrile, 3-methylbenzyl cyanide, m-tolylacetonitrile, m-methylphenylacetonitrile, m-xylyl cyanide, 3-methylbenzylcyanide, m-methylbenzyl cyanide, 3-methylphenylacetonitrile, 3-methylphenyl acetonitrile, 2-m-tolyl acetonitrile
PubChem CID 76279
IUPAC Name 2-(3-methylphenyl)acetonitrile
SMILES CC1=CC(=CC=C1)CC#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1055822 View Documents Alfa Aesar™
A1055822
100g Each for $271.42
 
Specifications

Chemical Identifiers

2947-60-6
131.178
WOJADIOTNFDWNQ-UHFFFAOYSA-N
76279
CC1=CC(=CC=C1)CC#N
C9H9N
MFCD00001914
2-3-methylphenyl acetonitrile, 3-methylbenzyl cyanide, m-tolylacetonitrile, m-methylphenylacetonitrile, m-xylyl cyanide, 3-methylbenzylcyanide, m-methylbenzyl cyanide, 3-methylphenylacetonitrile, 3-methylphenyl acetonitrile, 2-m-tolyl acetonitrile
2-(3-methylphenyl)acetonitrile

Specifications

>110°C (230°F)
240°C to 241°C
2947-60-6
MFCD00001914
1099644
WOJADIOTNFDWNQ-UHFFFAOYSA-N
2-(3-methylphenyl)acetonitrile
76279
98%
Almond-like
100g
C9H9N
UN3276
2-3-methylphenyl acetonitrile, 3-methylbenzyl cyanide, m-tolylacetonitrile, m-methylphenylacetonitrile, m-xylyl cyanide, 3-methylbenzylcyanide, m-methylbenzyl cyanide, 3-methylphenylacetonitrile, 3-methylphenyl acetonitrile, 2-m-tolyl acetonitrile
CC1=CC(=CC=C1)CC#N
131.178
131.18
m-Tolylacetonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.