missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

m-Tolunitrile, 99%, ACROS Organics™

Manufacturer:  Acros Organics AC158015000

 View more versions of this product

Catalog No. AC158015000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

m-Tolunitrile
Authentic
1.5246 to 1.5266 (20°C, 589nm)
500g
98.5
99%
CH3C6H4CN
09, 477
BOHCMQZJWOGWTA-UHFFFAOYSA-N
3-methylbenzonitrile
12104
Liquid
0.97g/mL
Glass bottle
Colorless to Yellow
620-22-4
100.0
C8H7N
MFCD00001808
m-tolunitrile, 3-cyanotoluene, benzonitrile, 3-methyl, m-cyanotoluene, 3-tolunitrile, m-toluonitrile, m-tolylnitrile, m-toluenenitrile, m-methylbenzonitrile, m-tolynitrile
CC1=CC=CC(=C1)C#N
117.151
117.15
≥98.5%

Chemical Identifiers

620-22-4
117.151
BOHCMQZJWOGWTA-UHFFFAOYSA-N
12104
CC1=CC=CC(=C1)C#N
C8H7N
MFCD00001808
m-tolunitrile, 3-cyanotoluene, benzonitrile, 3-methyl, m-cyanotoluene, 3-tolunitrile, m-toluonitrile, m-tolylnitrile, m-toluenenitrile, m-methylbenzonitrile, m-tolynitrile
3-methylbenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents