Please login to your online account to display your discounted pricing

Alfa Aesar™ m-Tolunitrile, 98+%

$163.91 - $163.91

Chemical Identifiers

Chemical Name or Material m-Tolunitrile
CAS 620-22-4
Molecular Formula C8H7N
Formula Weight 117.15g/mol
MDL Number MFCD00001808
Beilstein 507391
Synonym 3-Methylbenzonitrile; 3-Cyanotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1316530

 
alfa aesar™
A13165-30
250g Each for $163.91
Description & Specifications

Specifications

Odor Almond-like
Boiling Point 210°C
Melting Point −27°C
Flash Point 86°C (186°F)
Quantity 250g
Chemical Name or Material m-Tolunitrile
CAS 620-22-4
Assay ≥98%
Molecular Formula C8H7N
Formula Weight 117.15g/mol
MDL Number MFCD00001808
Beilstein 507391
Synonym 3-Methylbenzonitrile; 3-Cyanotoluene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS Number XV0525000
TSCA TSCA