missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ m-Toluenesulfonyl chloride, 98%

Manufacturer:  Alfa Aesar™ A1545803

 View more versions of this product

Catalog No. AAA1545803



Description

Description

Solubility
Reacts with water.

Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents.
Specifications

Specifications

m-Toluenesulfonyl chloride
Acrid
1g
C7H7ClO2S
UN3265
m-toluenesulfonyl chloride, 3-methylbenzene-1-sulfonyl chloride, 3-toluenesulfonyl chloride, 3-methyl-benzenesulfonyl chloride, 3-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 3-methyl, 3-toluenesulphonyl chloride, 3-methylphenylsulfonyl chloride, pubchem5624, m-tolylsulfonyl chloride
CC1=CC(=CC=C1)S(=O)(=O)Cl
190.641
190.65
107°C (224°F)
252°C to 253°C
1899-93-0
MFCD00051747
2087865
KFPMLWUKHQMEBU-UHFFFAOYSA-N
3-methylbenzenesulfonyl chloride
2734595
98%

Chemical Identifiers

1899-93-0
190.641
KFPMLWUKHQMEBU-UHFFFAOYSA-N
2734595
CC1=CC(=CC=C1)S(=O)(=O)Cl
C7H7ClO2S
MFCD00051747
m-toluenesulfonyl chloride, 3-methylbenzene-1-sulfonyl chloride, 3-toluenesulfonyl chloride, 3-methyl-benzenesulfonyl chloride, 3-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 3-methyl, 3-toluenesulphonyl chloride, 3-methylphenylsulfonyl chloride, pubchem5624, m-tolylsulfonyl chloride
3-methylbenzenesulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Documents