Learn More
m-Tolualdehyde, 98%, stabilized, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 138991000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
m-Tolualdehyde | |
620-23-5,123-31-9 | |
100.0 | |
199°C | |
Authentic | |
Glass bottle | |
1.5400 to 1.542 | |
MFCD00003374 | |
07, 296 | |
m-tolualdehyde, benzaldehyde, 3-methyl, m-methylbenzaldehyde, 3-tolylaldehyde, 3-tolualdehyde, m-tolyl aldehyde, m-toluylaldehyde, 3-methyl-benzaldehyd, unii-owh6650c4y, 3-methyl benzaldehyde | |
OVWYEQOVUDKZNU-UHFFFAOYSA-N | |
3-methylbenzaldehyde | |
0.1% hydroquinone | |
CHEBI:28476 | |
98% |
98% | |
97.5 | |
1.0190g/mL | |
78°C | |
97.5% min. (GC) | |
C8H8O | |
CH3C6H4CHO | |
100g | |
1.019 | |
Solubility in water: slightly soluble. Other solubilities: soluble in ethanol and ether | |
CC1=CC=CC(=C1)C=O | |
120.15 | |
12105 | |
120.15 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
May produce an allergic reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plen
GHS Signal Word: Warning
EINECSNumber : 210-632-0
TSCA : TSCA
Recommended Storage : Product darkens on storage
RUO â Research Use Only