Learn More
m-Tolualdehyde, 98%, stabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 138990250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
m-Tolualdehyde | |
620-23-5,123-31-9 | |
100.0 | |
1.0190g/mL | |
78°C | |
97.5% min. (GC) | |
C8H8O | |
CH3C6H4CHO | |
25 g | |
1.019 | |
Solubility in water: slightly soluble. Other solubilities: soluble in ethanol and ether | |
CC1=CC=CC(=C1)C=O | |
120.15 | |
12105 | |
120.15 | |
Liquid |
98% | |
97.5 | |
Brown to Yellow | |
199°C | |
Authentic | |
Glass bottle | |
1.5400 to 1.542 | |
MFCD00003374 | |
07, 296 | |
m-tolualdehyde, benzaldehyde, 3-methyl, m-methylbenzaldehyde, 3-tolylaldehyde, 3-tolualdehyde, m-tolyl aldehyde, m-toluylaldehyde, 3-methyl-benzaldehyd, unii-owh6650c4y, 3-methyl benzaldehyde | |
OVWYEQOVUDKZNU-UHFFFAOYSA-N | |
3-methylbenzaldehyde | |
0.1% hydroquinone | |
CHEBI:28476 | |
98% |
Chemical Identifiers
620-23-5 | |
120.15 | |
OVWYEQOVUDKZNU-UHFFFAOYSA-N | |
12105 | |
3-methylbenzaldehyde |
C8H8O | |
MFCD00003374 | |
m-tolualdehyde, benzaldehyde, 3-methyl, m-methylbenzaldehyde, 3-tolylaldehyde, 3-tolualdehyde, m-tolyl aldehyde, m-toluylaldehyde, 3-methyl-benzaldehyd, unii-owh6650c4y, 3-methyl benzaldehyde | |
CHEBI:28476 | |
CC1=CC=CC(=C1)C=O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
May produce an allergic reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plen
GHS Signal Word: Warning
EINECSNumber : 210-632-0
TSCA : TSCA
Recommended Storage : Product darkens on storage
RUO – Research Use Only