missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ m-Tolualdehyde, 97%, stab. with 0.1% hydroquinone

Manufacturer:  Alfa Aesar™ A1531322

 View more versions of this product

Catalog No. AAA1531322



Description

Description

Applications
m-Tolualdehyde is used as a fragrance agent in food and cosmetics. It is also used as a reagent in the preparation of a wide range of pharmaceutical compounds such as anti-inflammatory agents.

Solubility
Not miscible in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
Specifications

Specifications

m-Tolualdehyde
Almond-like
100g
620-23-5
MFCD00003374
m-tolualdehyde, benzaldehyde, 3-methyl, m-methylbenzaldehyde, 3-tolylaldehyde, 3-tolualdehyde, m-tolyl aldehyde, m-toluylaldehyde, 3-methyl-benzaldehyd, unii-owh6650c4y, 3-methyl benzaldehyde
CC1=CC=CC(=C1)C=O
120.151
CHEBI:28476
97%
78°C (172°F)
198°C to 200°C
stabilized with 0.1% hydroquinone
C8H8O
741964
OVWYEQOVUDKZNU-UHFFFAOYSA-N
3-methylbenzaldehyde
12105
120.15

Chemical Identifiers

620-23-5
120.151
OVWYEQOVUDKZNU-UHFFFAOYSA-N
12105
3-methylbenzaldehyde
C8H8O
MFCD00003374
m-tolualdehyde, benzaldehyde, 3-methyl, m-methylbenzaldehyde, 3-tolylaldehyde, 3-tolualdehyde, m-tolyl aldehyde, m-toluylaldehyde, 3-methyl-benzaldehyd, unii-owh6650c4y, 3-methyl benzaldehyde
CHEBI:28476
CC1=CC=CC(=C1)C=O
Safety and Handling

Safety and Handling

GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P301+P310-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : CU7033700

TSCA : TSCA

Documents