missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

m-Tolualdehyde 2,4-Dinitrophenylhydrazone, TCI America™

Manufacturer:  TCI America T14481G

 View more versions of this product

Catalog No. T14481G



Specifications

Specifications

m-Tolualdehyde 2,4-Dinitrophenylhydrazone
213°C
2880-05-9
MFCD00191585
m-tolualdehyde 2,4-dinitrophenylhydrazone, m-tolualdehyde, 2,4-dinitrophenyl hydrazone, benzaldehyde, 3-methyl-, 2,4-dinitrophenyl hydrazone, 3'-methylbenzaldehyde 2,4-dinitrophenylhydrazone, m-tolualdehyd-2,4-dinitrophenylhydrazone, m-tolualdehyde 2,4-dinitro-phenylhydrazone, 3-methylbenzaldehyde 2,4-dinitrophenyl hydrazone #, 1-2,4-dinitrophenyl-2-3-methylbenzylidene hydrazine, n-e-3-methylphenyl methylideneamino-2,4-dinitroaniline, m-tolualdehyde 2,4-dinitrophenylhydrazone, environmental standard
CC1=CC=CC(=C1)C=NNC2=C(C=C(C=C2)[N+](=O)[O-])[N+](=O)[O-]
300.27
300.27
≥98.0% (HPLC,T)
Undesignated
1g
C14H12N4O4
1325
PQBRXQUGVASQBU-UHFFFAOYSA-N
N-[(E)-(3-methylphenyl)methylideneamino]-2,4-dinitroaniline
9604108
Crystal-Powder at 20°C

Chemical Identifiers

2880-05-9
300.27
PQBRXQUGVASQBU-UHFFFAOYSA-N
9604108
CC1=CC=CC(=C1)C=NNC2=C(C=C(C=C2)[N+](=O)[O-])[N+](=O)[O-]
C14H12N4O4
MFCD00191585
m-tolualdehyde 2,4-dinitrophenylhydrazone, m-tolualdehyde, 2,4-dinitrophenyl hydrazone, benzaldehyde, 3-methyl-, 2,4-dinitrophenyl hydrazone, 3'-methylbenzaldehyde 2,4-dinitrophenylhydrazone, m-tolualdehyd-2,4-dinitrophenylhydrazone, m-tolualdehyde 2,4-dinitro-phenylhydrazone, 3-methylbenzaldehyde 2,4-dinitrophenyl hydrazone #, 1-2,4-dinitrophenyl-2-3-methylbenzylidene hydrazine, n-e-3-methylphenyl methylideneamino-2,4-dinitroaniline, m-tolualdehyde 2,4-dinitrophenylhydrazone, environmental standard
N-[(E)-(3-methylphenyl)methylideneamino]-2,4-dinitroaniline
Safety and Handling

Safety and Handling

TSCA : No

Documents