Please login to your online account to display your discounted pricing

Alfa Aesar™ m-Terphenyl, 99%

$44.15 - $44.15

Chemical Identifiers

Chemical Name or Material m-Terphenyl
CAS 92-06-8
Molecular Formula C18H14
Formula Weight 230.31
MDL Number MFCD00003059
Beilstein 1864778
Synonym 1,3-Diphenylbenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1486714

  SDS
alfa aesar™
L1486714
25g Each for $44.15
Description & Specifications

Specifications

Formula Weight 230.31
Chemical Name or Material m-Terphenyl
CAS 92-06-8
Assay 99%
Synonym 1,3-Diphenylbenzene
TSCA TSCA
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00003059
Beilstein 1864778
Molecular Formula C18H14
Flash Point 191°C (375°F)
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number WZ6470000
Boiling Point 363°C
Quantity 25g
Melting Point 86° to 88°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.