Learn More
m-Phenylenediamine dihydrochloride, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 216651000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
m-Phenylenediamine dihydrochloride | |
541-69-5 | |
100.0 | |
98.5% min. (Argentometry) | |
C6H8N2·2ClH | |
MFCD00012975 | |
13, 38 | |
benzene-1,3-diamine dihydrochloride, m-phenylenediamine dihydrochloride, 1,3-phenylenediamine dihydrochloride, 1,3-benzenediamine dihydrochloride, 1,3-benzenediamine, dihydrochloride, m-phenylenediamine hydrochloride, unii-se5rlo93tg, 1,3-diaminobenzene dihydrochloride, m-aminoaniline dihydrochloride, 3-aminoaniline dihydrochloride | |
SVTOYMIYCMHPIV-UHFFFAOYSA-N | |
benzene-1,3-diamine;dihydrochloride | |
10941 | |
99% |
99% | |
98.5 | |
Authentic | |
Glass bottle | |
C6H4(NH2)2·2HCl | |
100g | |
10, 7164 | |
Solubility in water: freely soluble. Other solubilities: soluble in alcohol | |
C1=CC(=CC(=C1)N)N.Cl.Cl | |
181.07 | |
181.07 |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
May cause an allergic skin reaction.
Causes serious eye irritation.
Toxic if inhaled.
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 208-790-0
RTECSNumber : SS9800000
TSCA : TSCA
RUO â Research Use Only