missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

m-Phenylenediamine, +99%, ACROS Organics™

$23.05 - $92.84

Chemical Identifiers

CAS 108-45-2
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Molecular Formula C6H8N2
Linear Formula C6H4(NH2)2
Formula Weight 108.14
MDL Number MFCD00007799
Synonym 1, 3-Diaminobenzene
Chemical Name or Material m-Phenylenediamine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC130560010 SDS Thermo Fisher Scientific
130560010
Glass bottle 1kg Each for $92.84
AC130560250 SDS Thermo Fisher Scientific
130560250
Glass bottle 25g Each for $23.05
AC130562500 SDS Thermo Fisher Scientific
130562500
Glass bottle 250g Each for $38.24
AC130565000 SDS Thermo Fisher Scientific
130565000
Glass bottle 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC130560050 SDS Thermo Fisher Scientific
AC130560050
Glass bottle 5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

108-45-2
100.0
C6H8N2
108.14
1, 3-Diaminobenzene
≥99.0% (p-phenylenediamine: <0.01%, o-phenylenediamine: <0.01%)
99.0
99+%
C6H4(NH2)2
MFCD00007799
m-Phenylenediamine

Specifications

Yellow to light tan or brown gray
1kg
99.0
Authentic
this product tends to darken during storage
99+%
C6H4(NH2)2
MFCD00007799
15, 7395
m-Phenylenediamine
63° to 65°C
108-45-2
100.0
Glass bottle
Pellets
C6H8N2
108.14
03, IV, 79
1, 3-Diaminobenzene
≥99.0% (p-phenylenediamine: <0.01%, o-phenylenediamine: <0.01%)

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger