Learn More
m-Phenylenediamine, 99+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 130560050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
m-Phenylenediamine | |
99.0 | |
282.0°C to 284.0°C | |
Authentic | |
Plastic drum | |
C6H4(NH2)2 | |
5kg | |
15, 7395 | |
Solubility in water: 350g/L (25°C). Other solubilities: soluble in alcohol,ether and chloroform | |
C1=CC(=CC(=C1)N)N | |
108.14 | |
CHEBI:8092 | |
99+% |
108-45-2 | |
100.0 | |
175°C | |
99% min. (GC) | |
C6H8N2 | |
MFCD00007799 | |
03, IV, 79 | |
m-phenylenediamine, 1,3-phenylenediamine, 1,3-diaminobenzene, 1,3-benzenediamine, m-diaminobenzene, 3-aminoaniline, m-aminoaniline, meta-phenylenediamine, developer c, 3-phenylenediamine | |
WZCQRUWWHSTZEM-UHFFFAOYSA-N | |
benzene-1,3-diamine | |
7935 | |
108.14 |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing genetic defects.
Very toxic to aquatic l
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim
GHS Signal Word: Danger
EINECSNumber : 203-584-7
Recommended Storage : this product tends to darken during storage
RUO â Research Use Only