Learn More
m-Phenetidine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 186720050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
m-Phenetidine | |
621-33-0 | |
100.0 | |
248.0°C | |
Authentic | |
Glass bottle | |
1.5650 to 1.5670 | |
MFCD00007785 | |
1.03 | |
WEZAHYDFZNTGKE-UHFFFAOYSA-N | |
3-ethoxyaniline | |
12120 | |
98% |
98% | |
97.5 | |
1.0300g/mL | |
>112°C | |
98% | |
C8H11NO | |
C2H5OC6H4NH2 | |
5mL | |
m-phenetidine, benzenamine, 3-ethoxy, m-ethoxyaniline, 3-ethoxybenzenamine, unii-xe9kkl972r, ccris 4696, xe9kkl972r, 3-ethoxyphenylamine, 3-ethoxy aniline, 3-ethoxy-phenylamine | |
CCOC1=CC=CC(N)=C1 | |
137.18 | |
137.18 |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:
GHS Signal Word: Danger
EINECSNumber : 210-680-2
RTECSNumber : SI5400000
TSCA : TSCA
RUO â Research Use Only