Learn More
m-Hydroxycinnamic acid, 99%, predominantly trans, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC121081000
Specifications
m-Hydroxycinnamic acid | |
Glass Bottle | |
189°C to 192°C | |
Predominantly trans | |
98.5 | |
99% | |
HOC6H4CH=CHCO2H | |
10, 294 | |
KKSDGJDHHZEWEP-SNAWJCMRSA-N | |
(E)-3-(3-hydroxyphenyl)prop-2-enoic acid | |
637541 | |
164.16 | |
99% |
Authentic | |
Beige | |
100g | |
14755-02-3 | |
100.0 | |
C9H8O3 | |
MFCD00004390 | |
2e-3-3-hydroxyphenyl prop-2-enoic acid, 3-3-hydroxyphenyl acrylic acid, 3-coumaric acid, 3-hydroxycinnamic acid, cinnamic acid, m-hydroxy, e-3-3-hydroxyphenyl acrylic acid, m-coumaric acid, m-hydroxycinnamic acid, trans-3-hydroxycinnamate, trans-3-hydroxycinnamic acid | |
C1=CC(=CC(=C1)O)C=CC(=O)O | |
164.16 | |
CHEBI:32357 | |
Fine Crystalline Powder |
Chemical Identifiers
14755-02-3 | |
164.16 | |
KKSDGJDHHZEWEP-SNAWJCMRSA-N | |
637541 | |
(E)-3-(3-hydroxyphenyl)prop-2-enoic acid |
C9H8O3 | |
MFCD00004390 | |
2e-3-3-hydroxyphenyl prop-2-enoic acid, 3-3-hydroxyphenyl acrylic acid, 3-coumaric acid, 3-hydroxycinnamic acid, cinnamic acid, m-hydroxy, e-3-3-hydroxyphenyl acrylic acid, m-coumaric acid, m-hydroxycinnamic acid, trans-3-hydroxycinnamate, trans-3-hydroxycinnamic acid | |
CHEBI:32357 | |
C1=CC(=CC(=C1)O)C=CC(=O)O |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning