Please login to your online account to display your discounted pricing

m-Ethyltoluene, 98%, Acros Organics

$108.46 - $108.46

Chemical Identifiers

Chemical Name or Material m-Ethyltoluene
Assay Percent Range 98%
Molecular Formula C9H12
Formula Weight 120.19
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC119000100

  SDS
thermo fisher scientific
AC119000100
Each for $108.46

AC119000250

  SDS
thermo fisher scientific
AC119000250
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 120.19
Quantity 10g
Chemical Name or Material m-Ethyltoluene
Assay Percent Range 98%
Molecular Formula C9H12
CAS 620-14-4