Please login to your online account to display your discounted pricing

m-Cresol Purple Sodium Salt, Reagent Grade, LabChem™

$347.74 - $347.74

Chemical Identifiers

Chemical Name or Material m-Cresol Purple Sodium Salt
CAS 62625-31-4
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula C21H17O5SNa
Formula Weight 404.42
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC134907

 
lab chem
LC134907
Amber Glass 10g Each for $347.74
Description & Specifications

Specifications

Formula Weight 404.42
Chemical Name or Material m-Cresol Purple Sodium Salt
Quantity 10g
EINECS Number 263-656-9
Purity Grade Reagent Grade
Physical Form Powder
Linear Formula C21H17O5SNa
CAS 62625-31-4
Identification Passes Test
Packaging Amber Glass
CAS Max % 100
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Solubility Soluble in water
Recommended Storage Room Temperature
Decomposition Information Sulfur compounds; Carbon monoxide; Carbon dioxide
Color Dark purple/black to orange
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature