Please login to your online account to display your discounted pricing

Alfa Aesar™ m-Cresol Purple sodium salt, 0.04% w/v aq. soln.

Chemical Identifiers

Chemical Name or Material m-Cresol Purple sodium salt
CAS 62625-31-4
Formula Weight 404.42
MDL Number MFCD00005871
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38691AE

  SDS
alfa aesar™
38691AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA38691AP

  SDS
alfa aesar™
38691AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA38691M0

  SDS
alfa aesar™
38691M0
6 x 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 404.42
Chemical Name or Material m-Cresol Purple sodium salt
Quantity 100mL
TSCA TSCA
Concentration 0.04% w/v aq. soln.
Physical Form Liquid
MDL Number MFCD00005871
CAS 62625-31-4