Please login to your online account to display your discounted pricing

m-Cresol Purple, 0.1% Aqueous, pH 1.2 to 2.8 Red to Yellow, pH 7.4 to 9.0 Yellow to Purple, Certified, LabChem™

$52.26 - $52.26

Chemical Identifiers

Chemical Name or Material m-Cresol Purple
CAS 7732-18-5
CAS Max % 99.9
CAS 62625-31-4
CAS Max % 0.1
Name Note 0.1% Aqueous, pH 1.2 to 2.8 Red to Yellow, pH 7.4 to 9.0 Yellow to Purple
Purity Grade Certified Grade
Linear Formula C21H17O5SNa
Formula Weight 404.42
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC135001

 
lab chem
LC135001
Poly Bottle 500mL Each for $52.26
Description & Specifications

Specifications

Formula Weight 404.42
Purity Grade Certified Grade
Physical Form Liquid
Solubility Soluble in water
Chemical Name or Material m-Cresol Purple
CAS 7732-18-5
CAS 62625-31-4
Linear Formula C21H17O5SNa
Recommended Storage Room Temperature
Decomposition Information Carbon monoxide; Carbon dioxide; Sulfur compounds
Specific Gravity 1g/mL
CAS Max % 99.9
Name Note 0.1% Aqueous, pH 1.2 to 2.8 Red to Yellow, pH 7.4 to 9.0 Yellow to Purple
Identification Passes Test
Color Dark, reddish-purple
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Quantity 500mL
CAS Max % 0.1
Packaging Poly Bottle
Density 1g/mL
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature