Alfa Aesar™ m-Carborane, 98%
Manufacturer:  Alfa Aesar™ 45606ME
 View more versions of this product
Catalog No. AA45606ME
Specifications
Specifications
m-Carborane
16986-24-6
MFCD00151574
JTWJUVSLJRLZFF-UHFFFAOYSA-N
2$l^{2},3$l^{2},4$l^{2},5$l^{2},6$l^{2},7$l^{2},8$l^{2},9$l^{2},11$l^{2},12$l^{2}-decaborabicyclo[8.1.1]dodecane
16212388
Powder
500mg
C2H2B10
m-carborane, 1,7-dicarbadodecaborane 12, m-carborane, 2,3,4,5,6,7,8,9,11,12-decaborabicyclo 8.1.1 dodecane
[B]1[B][B][B][B]C2[B]C([B]2)[B][B][B]1
134.138
144.25
98%
Chemical Identifiers
16986-24-6
134.138
JTWJUVSLJRLZFF-UHFFFAOYSA-N
16212388
[B]1[B][B][B][B]C2[B]C([B]2)[B][B][B]1
C2H2B10
MFCD00151574
m-carborane, 1,7-dicarbadodecaborane 12, m-carborane, 2,3,4,5,6,7,8,9,11,12-decaborabicyclo 8.1.1 dodecane
2$l^{2},3$l^{2},4$l^{2},5$l^{2},6$l^{2},7$l^{2},8$l^{2},9$l^{2},11$l^{2},12$l^{2}-decaborabicyclo[8.1.1]dodecane
Safety and Handling
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
TSCA : TSCA
SDS Documents