Learn More
m-Anisidine, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 104822500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
m-Anisidine | |
536-90-3 | |
250°C to 251°C | |
Authentic | |
Glass bottle | |
1.5800 to 1.582 | |
MFCD00007783 | |
13,404 | |
14,667 | |
Solubility in water: 18g/L (20°C). Other solubilities: soluble in acids,soluble in alcohol,diethyl ether,acetone and,benzene | |
COC1=CC=CC(=C1)N | |
123.15 | |
123.15 |
99% | |
1.1020g/mL | |
126°C | |
98.5% min. (GC) | |
C7H9NO | |
CH3OC6H4NH2 | |
250mL | |
1.102 | |
m-anisidine, 3-aminoanisole, m-methoxyaniline, 3-methoxybenzenamine, 3-anisidine, m-aminoanisole, benzenamine, 3-methoxy, m-anisylamine, 1-amino-3-methoxybenzene, unii-jxa144kx2i | |
NCBZRJODKRCREW-UHFFFAOYSA-N | |
3-methoxyaniline | |
10824 | |
99% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for
GHS Signal Word: Danger
EINECSNumber : 208-651-4
RTECSNumber : BZ5408000
TSCA : TSCA
RUO â Research Use Only