missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

m-Anisidine, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 104820050

 View more versions of this product

Catalog No. AC104820050



Description

Description

Specifications

Specifications

m-Anisidine
126°C
Glass bottle
1.102
Yellow to Brown
99%
98.5% min. (GC)
CH3OC6H4NH2
13,404
1-amino-3-methoxybenzene, 3-aminoanisole, 3-anisidine, 3-methoxybenzenamine, benzenamine, 3-methoxy, m-aminoanisole, m-anisidine, m-anisylamine, m-methoxyaniline, unii-jxa144kx2i
NCBZRJODKRCREW-UHFFFAOYSA-N
3-methoxyaniline
10824
Liquid
1.1020g/mL
Authentic
1.5800 to 1.582
250°C to 251°C
5mL
536-90-3
C7H9NO
MFCD00007783
14,667
Solubility in water: 18g/L (20°C). Other solubilities: soluble in acids,soluble in alcohol,diethyl ether,acetone and,benzene
COC1=CC=CC(=C1)N
123.15
123.15
99%

Chemical Identifiers

536-90-3
123.15
NCBZRJODKRCREW-UHFFFAOYSA-N
10824
COC1=CC=CC(=C1)N
C7H9NO
MFCD00007783
1-amino-3-methoxybenzene, 3-aminoanisole, 3-anisidine, 3-methoxybenzenamine, benzenamine, 3-methoxy, m-aminoanisole, m-anisidine, m-anisylamine, m-methoxyaniline, unii-jxa144kx2i
3-methoxyaniline
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for

GHS Signal Word: Danger

EINECSNumber : 208-651-4

RTECSNumber : BZ5408000

TSCA : TSCA

SDS Documents