Please login to your online account to display your discounted pricing

m-Anisidine, 99%, ACROS Organics™


Quantity 1L

Manufacturer: thermo fisher scientific  AC104820010

 View more versions of this product

Catalog No. AC104820010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 123.15
Assay ≥98.5%
Physical Form Liquid
Quantity 1L
Color Clear yellow to brown
TSCA TSCA
RTECS Number BZ5408000
EINECS Number 208-651-4
Density 1.1020g/mL
Packaging Glass bottle
Refractive Index 1.5800 to 1.5820 (20°C, 589nm)
Health Hazard 1 Danger
Name Note 99%
Molecular Formula C7H9NO
Assay Percent Range 99%
Linear Formula CH3OC6H4NH2
CAS 536-90-3
MDL Number MFCD00007783
Chemical Name or Material m-Anisidine
Infrared Spectrum Authentic
Synonym 3-Methoxyaniline
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material m-Anisidine
Name Note 99%
CAS 536-90-3
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H9NO
Linear Formula CH3OC6H4NH2
Formula Weight 123.15
MDL Number MFCD00007783
Synonym 3-Methoxyaniline
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.

SDS & Safety Documents