missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
m-Anisic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 152475000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
m-Anisic acid | |
103.0°C to 108.0°C | |
170.0°C to 172.0°C (10.0 mmHg) | |
97.5% min. (GC) | |
C8H8O3 | |
MFCD00002499 | |
10, 137 | |
XHQZJYCNDZAGLW-UHFFFAOYSA-N | |
3-methoxybenzoic acid | |
11461 | |
98% |
586-38-9 | |
Beige-Yellow to White | |
Authentic | |
Plastic bottle | |
CH3OC6H4CO2H | |
500 g | |
m-anisic acid, m-methoxybenzoic acid, benzoic acid, 3-methoxy, 3-anisic acid, meta-anisic acid, 3-methoxy-benzoic acid, unii-e2i36fh6qz, m-methylsalicylic acid, e2i36fh6qz, chembl22425 | |
COC1=CC=CC(=C1)C(=O)O | |
152.15 | |
152.15 | |
Powder |
Chemical Identifiers
586-38-9 | |
152.15 | |
XHQZJYCNDZAGLW-UHFFFAOYSA-N | |
11461 | |
COC1=CC=CC(=C1)C(=O)O |
C8H8O3 | |
MFCD00002499 | |
m-anisic acid, m-methoxybenzoic acid, benzoic acid, 3-methoxy, 3-anisic acid, meta-anisic acid, 3-methoxy-benzoic acid, unii-e2i36fh6qz, m-methylsalicylic acid, e2i36fh6qz, chembl22425 | |
3-methoxybenzoic acid |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 209-574-9
RUO – Research Use Only