missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

m-Anisic acid, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 152471000

 View more versions of this product

Catalog No. AC152471000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

m-Anisic acid
Plastic bottle
Beige-Yellow to White
100g
97.5% min. (GC)
CH3OC6H4CO2H
10, 137
XHQZJYCNDZAGLW-UHFFFAOYSA-N
3-methoxybenzoic acid
11461
Powder
Authentic
170.0°C to 172.0°C (10.0mmHg)
103.0°C to 108.0°C
586-38-9
C8H8O3
MFCD00002499
3-anisic acid, 3-methoxy-benzoic acid, benzoic acid, 3-methoxy, chembl22425, e2i36fh6qz, m-anisic acid, m-methoxybenzoic acid, m-methylsalicylic acid, meta-anisic acid, unii-e2i36fh6qz
COC1=CC=CC(=C1)C(=O)O
152.15
152.15
98%

Chemical Identifiers

586-38-9
152.15
XHQZJYCNDZAGLW-UHFFFAOYSA-N
11461
COC1=CC=CC(=C1)C(=O)O
C8H8O3
MFCD00002499
3-anisic acid, 3-methoxy-benzoic acid, benzoic acid, 3-methoxy, chembl22425, e2i36fh6qz, m-anisic acid, m-methoxybenzoic acid, m-methylsalicylic acid, meta-anisic acid, unii-e2i36fh6qz
3-methoxybenzoic acid
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 209-574-9

SDS Documents
Product Certifications