Please login to your online account to display your discounted pricing

M 344, Tocris

Histone deacetylase inhibitor

$82.94 - $746.48

Chemical Identifiers

Chemical Name or Material M 344
Assay Percent Range >98%
Molecular Formula C16H25N3O3
Formula Weight 307.39
Synonym 4-(Diethylamino)-N-[7-(hydroxyamino)-7-oxoheptyl]benzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

27711

 
r & d systems
RDS27711
1mg Each for $82.94

277110

 
r & d systems
RDS277110
10mg Each for $177.37

277150

 
r & d systems
RDS277150
50mg Each for $746.48
Description & Specifications

Specifications

Formula Weight 307.39
Quantity 1mg
CAS 251456-60-7
Molecular Formula C16H25N3O3
Chemical Name or Material M 344
Assay Percent Range >98%
Synonym 4-(Diethylamino)-N-[7-(hydroxyamino)-7-oxoheptyl]benzamide
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature