missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Lysing Matrix Z Bulk

Use for lysis of animal and plant tissue. MP Biomedicals Lysing Matrix Z Bulk contains 2.0mm diameter Yttria-stabilized Zirconium Oxide beads that are chemically inert and will not bind nucleic acids.

Manufacturer:  MP Biomedicals™ 116540438

 View more versions of this product

Catalog No. MP116540438



Description

Description

Used primarily for lysis of animal and plant tissue, including tougher stems, roots and seeds, and whole insects. The beads have high density and extremely high hardness, making this matrix ideal for cracking of very tough cell walls. Its smooth surface minimizes shearing, which minimizes degradation in applications such as RNA and protein isolation from very tough tissue.

Specifications

Specifications

Uniform Spheres
Store at Room Temperature (15–30°C).
Plants; Animal Tissues; Wood; Seeds; Soil; Clay; Bacteria; Ancient and Dried Samples
500g
1.5–2.0mm
Documents