missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals Lysing Matrix Z

$371.96 - $371.96

Specifications

For Use With (Application) Nucleic acid isolation and purification
For Use With (Equipment) FastPrep-24/120/96
Matrix Lysing matrix Z
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MP116961050 MP Biomedicals
116961050
50 x 2mL tubes Each for $371.96
Specifications Description & Specifications

Specifications

Nucleic acid isolation and purification
FastPrep-24/120/96
Lysing matrix Z