MP Biomedicals™ Lysing Matrix Y Bulk
Used primarily for lysis of gram positive and gram negative bacteria, fungal tissue and ores, yeasts, molds, filamentous algaes and biofilms.
Manufacturer:  MP Biomedicals™ 116540437
 View more versions of this product
Catalog No. MP116540437
Description
Description
  • Composed of 0.5mm diameter Yttria-Stabilized Zirconium Oxide beads
  • Y2O3 + ZrO2 has high density and has extremely high hardness making this matrix ideal for cracking of very tough cell walls, while it's smooth surface minimizes shearing
Specifications
Specifications
0.5mm Yttria-Stabilized Grinding Beads, Y Matrix
1kg
Documents